siRNA Details
| Gene Symbol: Lrrn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008516.4 | NP_032542.1 | 2 | 2959 | ||
| GenBank mRNA | AK139355.1 | - | 2 | 2917 | ||
| AK139965.1 | - | 2 | 2961 | |||
| AK160841.1 | - | 2 | 2838 | |||
| AK161885.1 | - | 1 | 1461 | |||
| AK173186.1 | - | 1 | 2204 | |||
| BC031122.1 | - | 1 | 2234 | 4972803 | ||
| BC058701.1 | - | 2 | 2892 | 6853964 | ||
| D45913.1 | - | 2 | 2905 | |||

Lrrn1
Genomic Map