siRNA Details
| Gene Symbol: GRK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005160.3 | NP_005151.2 | 21 | 3690 | ||
| XM_011529975.2 | XP_011528277.1 | 20 | 3400 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GRK3| Gene Symbol: GRK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005160.3 | NP_005151.2 | 21 | 3690 | ||
| XM_011529975.2 | XP_011528277.1 | 20 | 3400 | |||