siRNA Details
| Gene Symbol: Ly6f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008530.2 | NP_032556.1 | 4 | 604 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ly6f| Gene Symbol: Ly6f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008530.2 | NP_032556.1 | 4 | 604 | ||