siRNA Details
| Gene Symbol: Mcpt8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008572.1 | NP_032598.1 | 2 | 134 | ||
| GenBank mRNA | AK136485.1 | - | 2 | 154 | ||
| BC119342.1 | - | 2 | 210 | 8734091 | ||
| BC120607.1 | - | 2 | 207 | 40129530 | ||
| BC144934.1 | - | 2 | 210 | |||
| X78545.1 | - | 2 | 134 | |||

Mcpt8
Genomic Map