siRNA Details
| Gene Symbol: Mdm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF007110.1 | - | 3, 4 | 336 | ||
| AK017719.1 | - | 3, 4 | 310 | |||
| AK049576.1 | - | 3, 4 | 301 | |||
| AK049690.1 | - | 3, 4 | 305 | |||
| AK083349.1 | - | 4, 5 | 371 | |||
| AK139529.1 | - | 3, 4 | 246 | |||
| AK139535.1 | - | 3, 4 | 244 | |||
| AK161882.1 | - | 283 | ||||
| BC003750.1 | - | 3, 4 | 315 | 3599906 | ||
| CJ101093.1 | - | 3, 4 | 360 | |||
| EU568360.1 | - | 4, 5 | 367 | |||

Mdm4
Genomic Map