siRNA Details
| Gene Symbol: Myom2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008664.2 | NP_032690.2 | 2 | 183 | ||
| GenBank mRNA | AJ001038.1 | - | 2 | 183 | ||
| AK052683.1 | - | 2 | 330 | |||
| AK142174.1 | - | 2 | 159 | |||
| BC115722.1 | - | 2 | 174 | 40086642 | ||
| BC115834.1 | - | 2 | 173 | 40086646 | ||

Myom2
Genomic Map