siRNA Details
| Gene Symbol: OR2B2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033057.2 | NP_149046.2 | 1 | 673 | ||
| GenBank mRNA | BC136881.1 | - | 1 | 673 | 9020872 | |
| BC136901.1 | - | 1 | 673 | 9020892 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR2B2| Gene Symbol: OR2B2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033057.2 | NP_149046.2 | 1 | 673 | ||
| GenBank mRNA | BC136881.1 | - | 1 | 673 | 9020872 | |
| BC136901.1 | - | 1 | 673 | 9020892 | ||