siRNA Details
| Gene Symbol: DMRTB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033067.2 | NP_149056.1 | 4 | 1182 | ||
| GenBank mRNA | AJ291671.1 | - | 4 | 1109 | ||
| AK057273.1 | - | 4 | 1182 | |||
| AK302085.1 | - | 5 | 721 | |||
| BC029566.1 | - | 4 | 698 | 4836894 | ||

DMRTB1
Genomic Map