siRNA Details
| Gene Symbol: Anapc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008569.2 | NP_032595.2 | 2 | 464 | ||
| GenBank mRNA | AK052404.1 | - | 2 | 445 | ||
| AK053383.1 | - | 1 | 1537 | |||
| AK054305.1 | - | 2 | 446 | |||
| AK077847.1 | - | 2 | 465 | |||
| AK090134.1 | - | 2 | 447 | |||
| AK149762.1 | - | 2 | 436 | |||
| AK164804.1 | - | 2 | 450 | |||
| BC057201.1 | - | 2 | 423 | |||
| BC139002.1 | - | 2 | 445 | 8862024 | ||
| BC145492.1 | - | 2 | 445 | 9054055 | ||
| X80169.1 | - | 2 | 449 | |||

Anapc1
Genomic Map