siRNA Details
| Gene Symbol: Mcpt8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008572.1 | NP_032598.1 | 4 | 414 | ||
| GenBank mRNA | AK136485.1 | - | 4 | 434 | ||
| BC119342.1 | - | 4 | 490 | 8734091 | ||
| BC120607.1 | - | 4 | 487 | 40129530 | ||
| BC144934.1 | - | 4 | 576 | |||
| X78545.1 | - | 4 | 414 | |||

Mcpt8
Genomic Map