siRNA Details
| Gene Symbol: Smcp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008574.3 | NP_032600.2 | 1, 2 | 204 | ||
| GenBank mRNA | AK077172.1 | - | 1, 2 | 175 | ||
| BC055104.1 | - | 1, 2 | 174 | 6773089 | ||
| BC061167.1 | - | 1, 2 | 154 | 6771333 | ||
| BU936528.1 | - | 1, 2 | 207 | |||
| M29603.1 | - | 1, 2 | 177 | |||
| S49659.1 | - | 1, 2 | 174 | |||
| S49660.1 | - | 324 | ||||

Smcp
Genomic Map