siRNA Details
| Gene Symbol: Mfng | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008595.2 | NP_032621.1 | 1, 2 | 429 | ||
| GenBank mRNA | AF015769.1 | - | 1, 2 | 256 | ||
| AK143523.1 | - | 1, 2 | 463 | |||
| BC010983.1 | - | 1, 2 | 451 | 3980692 | ||
| U94349.1 | - | 1, 2 | 429 | |||

Mfng
Genomic Map