siRNA Details
| Gene Symbol: Zfp239 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001792.1 | NP_001001792.1 | 4 | 1951 | ||
| NM_008616.2 | NP_032642.2 | 3 | 1771 | |||
| GenBank mRNA | AK078513.1 | - | 3 | 1765 | ||
| AK133328.1 | - | 1610 | ||||
| BC003315.1 | - | 1 | 705 | 3155528 | ||
| BC080674.1 | - | 1 | 1588 | |||
| M32057.1 | - | 1 | 1478 | |||

Zfp239
Genomic Map