siRNA Details
| Gene Symbol: Mybpc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008653.2 | NP_032679.2 | 2 | 318 | ||
| GenBank mRNA | AF097333.1 | - | 2 | 269 | ||
| AK146661.1 | - | 2 | 319 | |||
| AK146886.1 | - | 2 | 319 | |||
| AK169302.1 | - | 2 | 299 | |||
| AK169306.1 | - | 2 | 299 | |||
| BC094945.1 | - | 2 | 318 | |||
| BC131672.1 | - | 2 | 294 | |||
| BC154408.1 | - | 2 | 339 | 40110890 | ||

Mybpc3
Genomic Map