siRNA Details
| Gene Symbol: Smcp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008574.3 | NP_032600.2 | 2 | 830 | ||
| GenBank mRNA | AK077172.1 | - | 2 | 801 | ||
| BC055104.1 | - | 2 | 800 | 6773089 | ||
| BC061167.1 | - | 2 | 780 | 6771333 | ||
| BY456161.1 | - | 1 | 373 | |||
| M29603.1 | - | 2 | 803 | |||

Smcp
Genomic Map