siRNA Details
| Gene Symbol: Met | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008591.2 | NP_032617.2 | 13 | 2974 | ||
| GenBank mRNA | BC117969.1 | - | 11 | 2618 | ||
| BC117970.1 | - | 11 | 2614 | |||
| CK332973.1 | - | 2 | 90 | |||
| Y00671.1 | - | 11 | 2619 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Met| Gene Symbol: Met | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008591.2 | NP_032617.2 | 13 | 2974 | ||
| GenBank mRNA | BC117969.1 | - | 11 | 2618 | ||
| BC117970.1 | - | 11 | 2614 | |||
| CK332973.1 | - | 2 | 90 | |||
| Y00671.1 | - | 11 | 2619 | |||