siRNA Details
| Gene Symbol: Mmp15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008609.3 | NP_032635.1 | 7 | 1806 | ||
| GenBank mRNA | AY622975.1 | - | 2 | 213 | ||
| BC047278.1 | - | 7 | 1806 | 6390113 | ||
| BC057952.1 | - | 7 | 1376 | 5342023 | ||
| D86332.1 | - | 7 | 1302 | |||

Mmp15
Genomic Map