siRNA Details
| Gene Symbol: Map7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB098611.1 | - | 9 | 946 | ||
| Y15197.1 | - | 9 | 946 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Map7| Gene Symbol: Map7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB098611.1 | - | 9 | 946 | ||
| Y15197.1 | - | 9 | 946 | |||