siRNA Details
| Gene Symbol: Myom2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008664.2 | NP_032690.2 | 3 | 349 | ||
| GenBank mRNA | AJ001038.1 | - | 3 | 349 | ||
| AK052683.1 | - | 3 | 496 | |||
| AK142174.1 | - | 3 | 325 | |||
| BC115722.1 | - | 3 | 340 | 40086642 | ||
| BC115834.1 | - | 3 | 339 | 40086646 | ||

Myom2
Genomic Map