siRNA Details
| Gene Symbol: Nab1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008667.3 | NP_032693.2 | 8 | 1771 | ||
| GenBank mRNA | AK018122.1 | - | 2 | 183 | ||
| BC005627.1 | - | 8 | 1771 | 3155795 | ||
| BC016886.1 | - | 8 | 1694 | 4236565 | ||
| U47008.1 | - | 8 | 1650 | |||

Nab1
Genomic Map