siRNA Details
Gene Symbol: Map4k4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252200.1 | NP_001239129.1 | 2, 3 | 448 | ||
NM_001252201.1 | NP_001239130.1 | 2, 3 | 448 | |||
NM_001252202.1 | NP_001239131.1 | 2, 3 | 448 | |||
NM_008696.2 | NP_032722.2 | 2, 3 | 448 | |||
GenBank mRNA | AK129191.1 | - | 2, 3 | 185 | ||
BC141109.1 | - | 2, 3 | 182 | 9055721 | ||
BC145390.1 | - | 2, 3 | 182 | 9053948 | ||
BC145391.1 | - | 2, 3 | 181 | 9053949 | ||
CJ142598.1 | - | 2, 3 | 455 | |||
U88984.1 | - | 2, 3 | 131 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
