siRNA Details
| Gene Symbol: Mycn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008709.3 | NP_032735.3 | 2 | 1067 | ||
| GenBank mRNA | AK144126.1 | - | 2 | 1067 | ||
| AK146696.1 | - | 2 | 1068 | |||
| AK146913.1 | - | 2 | 1065 | |||
| BC005453.1 | - | 2 | 951 | 3495446 | ||
| BC049783.1 | - | 1 | 996 | 6517457 | ||
| M36277.1 | - | 843 | ||||

Mycn
Genomic Map