siRNA Details
| Gene Symbol: Olfr15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008762.2 | NP_032788.2 | 1 | 548 | ||
| GenBank mRNA | BC100679.1 | - | 1 | 546 | ||
| BC100680.1 | - | 1 | 547 | |||
| BC100685.1 | - | 1 | 547 | |||
| BC115838.1 | - | 1 | 547 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr15| Gene Symbol: Olfr15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008762.2 | NP_032788.2 | 1 | 548 | ||
| GenBank mRNA | BC100679.1 | - | 1 | 546 | ||
| BC100680.1 | - | 1 | 547 | |||
| BC100685.1 | - | 1 | 547 | |||
| BC115838.1 | - | 1 | 547 | |||