siRNA Details
| Gene Symbol: HRH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022304.2 | NP_071640.1 | 1 | 3051 | ||
| XM_006714865.3 | XP_006714928.1 | 3 | 2089 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HRH2| Gene Symbol: HRH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022304.2 | NP_071640.1 | 1 | 3051 | ||
| XM_006714865.3 | XP_006714928.1 | 3 | 2089 | |||
Set Membership: |
|