siRNA Details
| Gene Symbol: Ncoa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB219976.1 | - | 2 | 131 | ||
| AF000582.1 | - | 4 | 282 | |||
| AK038781.1 | - | 4 | 455 | |||
| AK048972.1 | - | 4 | 300 | |||
| AK052117.1 | - | 4 | 353 | |||
| AK087443.1 | - | 5 | 361 | |||
| BC053387.1 | - | 4 | 476 | 30054358 | ||
| U39060.1 | - | 4 | 334 | |||

Ncoa2
Genomic Map