siRNA Details
| Gene Symbol: Notch3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008716.2 | NP_032742.1 | 5, 6 | 872 | ||
| GenBank mRNA | X74760.1 | - | 5, 6 | 872 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Notch3| Gene Symbol: Notch3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008716.2 | NP_032742.1 | 5, 6 | 872 | ||
| GenBank mRNA | X74760.1 | - | 5, 6 | 872 | ||