siRNA Details
| Gene Symbol: Nsf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008740.4 | NP_032766.2 | 6 | 533 | ||
| GenBank mRNA | AK042341.1 | - | 7 | 551 | ||
| AK049281.1 | - | 6 | 503 | |||
| AK051430.1 | - | 6 | 502 | |||
| AK085086.1 | - | 6 | 487 | |||
| AK141986.1 | - | 6 | 503 | |||
| AK153905.1 | - | 6 | 502 | |||
| BC006627.1 | - | 6 | 538 | 3968733 | ||
| BC019167.1 | - | 6 | 486 | 4506351 | ||
| U10120.1 | - | 6 | 491 | |||

Nsf
Genomic Map