siRNA Details
| Gene Symbol: Ogn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008760.4 | NP_032786.1 | 7 | 902 | ||
| GenBank mRNA | AK028300.1 | - | 7 | 929 | ||
| AK053650.1 | - | 7 | 903 | |||
| AK075638.1 | - | 6 | 860 | |||
| AK160760.1 | - | 7 | 924 | |||
| AK163772.1 | - | 6 | 854 | |||
| BC021939.1 | - | 6 | 805 | 5067073 | ||
| D31951.1 | - | 6 | 815 | |||

Ogn
Genomic Map