siRNA Details
| Gene Symbol: Slc22a18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042760.1 | NP_001036225.1 | 7, 8 | 980 | ||
| NM_008767.2 | NP_032793.2 | 7, 8 | 940 | |||
| GenBank mRNA | AB012084.1 | - | 7, 8 | 980 | ||
| AF028739.1 | - | 7, 8 | 896 | |||
| AF037065.1 | - | 7, 8 | 908 | |||
| AK002460.1 | - | 7, 8 | 922 | |||
| BC003734.1 | - | 7, 8 | 886 | 3599736 | ||
| BC015078.1 | - | 6, 7 | 827 | 4224844 | ||
| BE865039.1 | - | 4, 5 | 447 | |||

Slc22a18
Genomic Map