siRNA Details
| Gene Symbol: GPR87 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023915.3 | NP_076404.3 | 3 | 1214 | ||
| GenBank mRNA | AB031548.1 | - | 3 | 1146 | ||
| AF086432.1 | - | 1 | 289 | |||
| AF237763.1 | - | 3 | 1102 | |||
| AF345566.1 | - | 1 | 892 | |||
| AF411114.1 | - | 2 | 853 | |||
| AK027784.1 | - | 3 | 1214 | |||
| BC009540.1 | - | 1 | 660 | |||
| BC112941.1 | - | 3 | 1123 | 4500655 | ||
| BC117565.1 | - | 3 | 1123 | 4521950 | ||
| BC132888.1 | - | 3 | 1180 | 40146910 | ||
| BC132890.1 | - | 3 | 1180 | 40146912 | ||
| EU883576.1 | - | 2 | 853 | |||
More Information
Set Membership: |
|

GPR87
Genomic Map