siRNA Details
| Gene Symbol: P2ry2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005013.2 | - | 3 | 1521 | ||
| AK017378.1 | - | 3 | 1528 | |||
| AK155521.1 | - | 2 | 1373 | |||
| AK155562.1 | - | 2 | 1373 | |||
| AK156185.1 | - | 3 | 1325 | |||
| BC006613.1 | - | 3 | 1515 | 3967332 | ||
| L14751.1 | - | 2 | 1314 | |||
| S83099.1 | - | 1 | 1045 | |||

P2ry2
Genomic Map