siRNA Details
| Gene Symbol: Map4k4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252200.1 | NP_001239129.1 | 6, 7 | 823 | ||
| NM_001252201.1 | NP_001239130.1 | 6, 7 | 823 | |||
| NM_001252202.1 | NP_001239131.1 | 6, 7 | 823 | |||
| NM_008696.2 | NP_032722.2 | 6, 7 | 823 | |||
| GenBank mRNA | AK129191.1 | - | 6, 7 | 560 | ||
| BC056650.1 | - | 6, 7 | 566 | |||
| BC141109.1 | - | 6, 7 | 557 | 9055721 | ||
| BC145390.1 | - | 6, 7 | 557 | 9053948 | ||
| BC145391.1 | - | 6, 7 | 556 | 9053949 | ||
| CF531764.1 | - | 4, 5 | 381 | |||
| U88984.1 | - | 6, 7 | 503 | |||

Map4k4
Genomic Map