siRNA Details
| Gene Symbol: Npr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039181.1 | NP_001034270.1 | 7, 8 | 1539 | ||
| NM_001286395.1 | NP_001273324.1 | 8, 9 | 1155 | |||
| NM_008728.2 | NP_032754.2 | 7, 8 | 1542 | |||
| GenBank mRNA | AA763613.1 | - | 2, 3 | 222 | ||
| AK079196.1 | - | 8, 9 | 1072 | |||
| AK166535.1 | - | 6, 7 | 770 | |||
| BC055897.1 | - | 7, 8 | 1539 | 4019152 | ||
| BQ831432.1 | - | 4, 5 | 390 | |||
| D78175.1 | - | 7, 8 | 1507 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_012868, XM_006232045, L27339, X78595, X75605.

Npr3
Genomic Map