siRNA Details
| Gene Symbol: Ogn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008760.4 | NP_032786.1 | 7 | 2469 | ||
| GenBank mRNA | AK028300.1 | - | 7 | 2496 | ||
| AK053650.1 | - | 7 | 2470 | |||
| AK163772.1 | - | 6 | 2421 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ogn| Gene Symbol: Ogn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008760.4 | NP_032786.1 | 7 | 2469 | ||
| GenBank mRNA | AK028300.1 | - | 7 | 2496 | ||
| AK053650.1 | - | 7 | 2470 | |||
| AK163772.1 | - | 6 | 2421 | |||