siRNA Details
Gene Symbol: P2ry1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282016.1 | NP_001268945.1 | 2 | 1879 | ||
NM_008772.5 | NP_032798.1 | 2 | 2154 | |||
GenBank mRNA | AK030759.1 | - | 2 | 1868 | ||
AK036611.1 | - | 2 | 2145 | |||
BC138634.1 | - | 2 | 2015 | 8861654 | ||
BC138635.1 | - | 2 | 2015 | 8861655 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
