siRNA Details
| Gene Symbol: Pde8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008803.2 | NP_032829.1 | 1, 2 | 293 | ||
| GenBank mRNA | AF067806.1 | - | 359 | |||
| AK148238.1 | - | 502 | ||||
| AK149561.1 | - | 1, 2 | 494 | |||
| BC125578.1 | - | 1, 2 | 293 | 40130093 | ||
| BC132145.1 | - | 1, 2 | 293 | 40130422 | ||

Pde8a
Genomic Map