siRNA Details
| Gene Symbol: Pdx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008814.3 | NP_032840.1 | 2 | 551 | ||
| GenBank mRNA | AK020261.1 | - | 556 | |||
| BC103572.1 | - | 2 | 551 | 40045668 | ||
| BC103581.1 | - | 2 | 551 | 40045664 | ||
| BC103582.1 | - | 2 | 551 | 40045666 | ||
| BC105642.1 | - | 2 | 551 | 40045665 | ||
| X74342.1 | - | 2 | 722 | |||

Pdx1
Genomic Map