siRNA Details
| Gene Symbol: Pip4k2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008845.4 | NP_032871.3 | 2 | 353 | ||
| GenBank mRNA | AB009615.1 | - | 2 | 278 | ||
| AK012196.1 | - | 2 | 356 | |||
| AK040558.1 | - | 2 | 243 | |||
| BC011097.1 | - | 2 | 300 | 3672732 | ||

Pip4k2a
Genomic Map