siRNA Details
| Gene Symbol: Prkcz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008860.3 | NP_032886.2 | 2 | 328 | ||
| GenBank mRNA | AK165750.1 | - | 2 | 243 | ||
| BC139761.1 | - | 2 | 145 | 40109623 | ||
| BY073558.1 | - | 2 | 292 | |||
| CJ169725.1 | - | 2 | 329 | |||
| CK339309.1 | - | 2 | 77 | |||
| M94632.1 | - | 2 | 145 | |||

Prkcz
Genomic Map