siRNA Details
| Gene Symbol: PBX4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025245.2 | NP_079521.1 | 3 | 635 | ||
| XM_006722911.3 | XP_006722974.1 | 2 | 216 | |||
| XM_011528320.2 | XP_011526622.1 | 4 | 681 | |||
| XM_011528322.2 | XP_011526624.1 | 2 | 270 | |||
| XM_017027326.1 | XP_016882815.1 | 4 | 316 | |||
| XM_017027327.1 | XP_016882816.1 | 4 | 297 | |||
| XM_017027328.1 | XP_016882817.1 | 4 | 681 | |||
| GenBank mRNA | AJ300182.1 | - | 2 | 216 | ||
| AK097427.1 | - | 3 | 372 | |||
| BC141859.1 | - | 3 | 635 | 8860384 | ||
| BG683004.1 | - | 3 | 350 | |||

PBX4
Genomic Map