siRNA Details
| Gene Symbol: Cdk18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008795.2 | NP_032821.1 | 11 | 1281 | ||
| GenBank mRNA | AK004998.1 | - | 11 | 1225 | ||
| AK171569.1 | - | 11 | 1062 | |||
| BC119617.1 | - | 12 | 1157 | 40061728 | ||
| BC119618.1 | - | 6 | 548 | |||
| X69026.1 | - | 11 | 1298 | |||

Cdk18
Genomic Map