siRNA Details
| Gene Symbol: Peg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008817.2 | NP_032843.2 | 9 | 1233 | ||
| GenBank mRNA | AB003040.1 | - | 9 | 1230 | ||
| AF038939.1 | - | 6 | 884 | |||
| AK044602.1 | - | 10 | 1297 | |||
| AK122238.1 | - | 9 | 1212 | |||
| AK141217.1 | - | 9 | 1234 | |||
| AK163845.1 | - | 9 | 1236 | |||
| BC048778.1 | - | 4 | 610 | 6308481 | ||
| BC072661.1 | - | 9 | 1221 | 30543809 | ||
| BC085183.1 | - | 9 | 1218 | 30663888 | ||
| BC089344.1 | - | 1 | 313 | |||

Peg3
Genomic Map