siRNA Details
| Gene Symbol: Prkcz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008860.3 | NP_032886.2 | 2, 3 | 407 | ||
| GenBank mRNA | AK165750.1 | - | 2, 3 | 322 | ||
| BC139761.1 | - | 2, 3 | 224 | 40109623 | ||
| CK339309.1 | - | 2, 3 | 156 | |||
| M94632.1 | - | 2, 3 | 224 | |||

Prkcz
Genomic Map