siRNA Details
| Gene Symbol: Pcnt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282992.1 | NP_001269921.1 | 6 | 1085 | ||
| NM_008787.3 | NP_032813.3 | 6 | 1085 | |||
| GenBank mRNA | AB207233.1 | - | 6 | 959 | ||
| AK145315.1 | - | 7 | 906 | |||
| BC085244.1 | - | 6 | 909 | |||
| BC100380.1 | - | 6 | 880 | |||
| BU518252.1 | - | 606 | ||||

Pcnt
Genomic Map