siRNA Details
| Gene Symbol: Phxr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | X12808.1 | - | 959 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Phxr2| Gene Symbol: Phxr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | X12808.1 | - | 959 | |||