siRNA Details
Gene Symbol: Pou3f4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008901.1 | NP_032927.1 | 1 | 551 | ||
GenBank mRNA | AK158302.1 | - | 1 | 584 | ||
BC052012.1 | - | 1 | 599 | |||
BC086670.1 | - | 562 | ||||
BC138657.1 | - | 1 | 626 | 8861677 | ||
BC145919.1 | - | 1 | 626 | 40131222 | ||
X66603.1 | - | 594 |
