siRNA Details
| Gene Symbol: Psma2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008944.2 | NP_032970.2 | 3 | 220 | ||
| GenBank mRNA | AF099733.1 | - | 3 | 187 | ||
| AK012119.1 | - | 3 | 202 | |||
| AK012990.1 | - | 3 | 209 | |||
| AK035578.1 | - | 209 | ||||
| AK136115.1 | - | 3 | 222 | |||
| AK151883.1 | - | 3 | 211 | |||
| AK152862.1 | - | 3 | 211 | |||
| AK154011.1 | - | 3 | 204 | |||
| AK167752.1 | - | 3 | 211 | |||
| BC026768.1 | - | 3 | 193 | 3156907 | ||
| CJ052982.1 | - | 3 | 222 | |||
| X70303.1 | - | 3 | 264 | |||

Psma2
Genomic Map