siRNA Details
| Gene Symbol: Pten | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008960.2 | NP_032986.1 | 2 | 1006 | ||
| GenBank mRNA | AK076980.1 | - | 2 | 964 | ||
| AK088717.1 | - | 2 | 985 | |||
| AK164302.1 | - | 2 | 989 | |||
| AK208703.1 | - | 2 | 238 | |||
| BC021445.1 | - | 2 | 355 | 5038272 | ||
| U92437.1 | - | 2 | 1086 | |||

Pten
Genomic Map