siRNA Details
| Gene Symbol: Ptpn14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008976.2 | NP_033002.2 | 3, 4 | 877 | ||
| GenBank mRNA | AF170902.1 | - | 2, 3 | 444 | ||
| AF170903.1 | - | 2, 3 | 444 | |||
| AK028783.1 | - | 3, 4 | 841 | |||
| AK037646.1 | - | 3, 4 | 878 | |||
| AK086524.1 | - | 3, 4 | 871 | |||
| AK156480.1 | - | 3, 4 | 623 | |||
| D31842.1 | - | 2, 3 | 474 | |||

Ptpn14
Genomic Map