siRNA Details
| Gene Symbol: Ptpn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127177.1 | NP_001120649.1 | 3 | 353 | ||
| NM_008977.3 | NP_033003.1 | 3 | 353 | |||
| GenBank mRNA | AK014528.1 | - | 3 | 321 | ||
| AK076072.1 | - | 3 | 306 | |||
| AK132013.1 | - | 3 | 316 | |||
| BC008269.1 | - | 3 | 327 | 3602983 | ||
| CA560046.1 | - | 3 | 353 | |||
| M80739.1 | - | 3 | 308 | |||
| M81477.1 | - | 3 | 308 | |||
| S52655.1 | - | 3 | 303 | |||

Ptpn2
Genomic Map